Assay Details
| Gene Symbol: Ctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013256.2 | NP_001013274.2 | 20 - 21 | 3428 | 64 | |
| NM_001143790.1 | NP_001137262.1 | 20 - 21 | 3425 | 64 | ||
| NM_001281464.1 | NP_001268393.1 | 20 - 21 | 3296 | 64 | ||
| NM_001281465.1 | NP_001268394.1 | 20 - 21 | 3412 | 64 | ||
| GenBank mRNA | AK038651.1 | - | 20 - 21 | 3212 | 64 | |
| AK044055.1 | - | 20 - 21 | 3207 | 64 | ||
| AK145919.1 | - | 20 - 21 | 3243 | 64 | ||
| AK165906.1 | - | 20 - 21 | 3228 | 64 | ||
| BC014687.1 | - | 11 - 12 | 1596 | 64 | ||
| BC060629.1 | - | 16 - 17 | 2478 | 6847619 | 64 | |
| BC066078.1 | - | 20 - 21 | 3239 | 64 | ||
| BC072625.1 | - | 20 - 21 | 3253 | 64 | ||
| BC147495.1 | - | 20 - 21 | 3226 | 9056325 | 64 | |
| BC147497.1 | - | 20 - 21 | 3226 | 9056354 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ctc1
Genomic Map