Assay Details
| Gene Symbol: Snx15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026912.1 | NP_081188.1 | 1 - 2 | 164 | 70 | |
| GenBank mRNA | BC016091.1 | - | 1 - 2 | 164 | 4503873 | 70 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90327 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702266 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 671969 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 51927 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115755 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932869 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932882 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932883 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932878 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932847 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932832 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932867 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932873 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932877 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932863 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932830 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932880 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932864 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932871 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932886 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932874 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932844 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932836 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932850 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932849 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932831 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932884 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932865 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932833 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932888 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932845 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932842 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932876 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932854 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932840 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932846 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932861 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932843 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932889 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31932838 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03544216 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115755 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09095474 | - | - |
| Whole Mouse Genome G4122A | A_52_P462106 | - | - |
| Whole Mouse Genome G4122A | A_52_P3283 | - | - |
| Whole Mouse Genome G4122A | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | AK197572.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | BC016091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | NM_026912.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P205968 | AK005338.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115050 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03544216 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115755 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14281554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14421825 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14281553 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14281551 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14261166 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14281557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932863 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932909 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932873 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932818 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31932767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14281810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932895 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932867 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14281804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932858 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932849 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932846 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932831 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932851 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14384353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14281806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14142499 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932836 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14157274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932890 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14281554 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932874 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932888 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932834 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14402717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932848 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14421825 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14402716 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14374430 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14168569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932871 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932859 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14402713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932876 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932869 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932843 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31932861 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465386 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465380 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465387 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465385 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465383 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465384 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465381 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465382 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424337_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465379 | - | - |
| Mouse Genome 430 2.0 Array | 1424337_at | - | - |
| Mouse Genome 430 2.0 Array | 1456649_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424337_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1456649_at | - | - |
| Mouse Genome 430 2.0 Array | 1458021_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1458021_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69024_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69024_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900009269.mm.1 | NM_026912.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009269.mm.1 | NM_026912.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009269.mm.1 | TC1900001003.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009283.mm.1 | NM_026912.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009283.mm.1 | NM_026912.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009283.mm.1 | TC1900001003.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900009277.mm.1 | NM_026912.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009277.mm.1 | NM_026912.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009277.mm.1 | TC1900001003.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424337_PM_at | - | - |
| Mouse Expression Set 430 | 1424337_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424337_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424337_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424337_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424337_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 885310 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1240348 | - | - |
| MouseWG-6 v2 Expression BeadChip | 6770082 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6770082 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1240348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006770082 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001240348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2628871 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2740337 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2628871 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2740337 | - | - |
| mouse-6 v1.1 (Illumina) | scl53529.8.1_144-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000539.1_19-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6770082 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1240348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000539.1_19-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53529.8.1_144-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1240348 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6770082 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2740337 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2894450 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2894457 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2740337 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2894457 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2894450 | - | - |

Snx15
Genomic Map