Assay Details
| Gene Symbol: Tram1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028304.1 | - | 6 - 7 | 723 | 103 | |
| AK088814.1 | - | 6 - 7 | 713 | 103 | ||
| AK142658.1 | - | 6 - 7 | 713 | 103 | ||
| AK145291.1 | - | 6 - 7 | 708 | 103 | ||
| AK146207.1 | - | 6 - 7 | 712 | 103 | ||
| AK150551.1 | - | 6 - 7 | 719 | 103 | ||
| AK150777.1 | - | 6 - 7 | 677 | 103 | ||
| AK151240.1 | - | 6 - 7 | 676 | 103 | ||
| AK151409.1 | - | 6 - 7 | 719 | 103 | ||
| AK151664.1 | - | 6 - 7 | 719 | 103 | ||
| AK152338.1 | - | 6 - 7 | 719 | 103 | ||
| AK152343.1 | - | 6 - 7 | 678 | 103 | ||
| AK152991.1 | - | 6 - 7 | 678 | 103 | ||
| AK153379.1 | - | 6 - 7 | 719 | 103 | ||
| AK155905.1 | - | 6 - 7 | 713 | 103 | ||
| AK159203.1 | - | 6 - 7 | 713 | 103 | ||
| AK159350.1 | - | 6 - 7 | 713 | 103 | ||
| AK159878.1 | - | 6 - 7 | 722 | 103 | ||
| AK168708.1 | - | 6 - 7 | 655 | 103 | ||
| AY029764.1 | - | 6 - 7 | 605 | 103 | ||
| BC012401.1 | - | 6 - 7 | 702 | 3967323 | 103 | |
| CT010238.1 | - | 6 - 7 | 570 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tram1
Genomic Map