Assay Details
| Gene Symbol: Dzip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028258.3 | NP_082534.2 | 13 - 14 | 1986 | 79 | |
| GenBank mRNA | AK028787.1 | - | 14 - 15 | 2141 | 79 | |
| BC099950.1 | - | 13 - 14 | 1986 | 30094307 | 79 | |
| BC138218.1 | - | 12 - 13 | 1915 | 8861237 | 79 | |
More Information
Set Membership: |
|

Dzip1l
Genomic Map