Assay Details
| Gene Symbol: Dzip1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028258.3 | NP_082534.2 | 4 - 5 | 855 | 97 | |
| GenBank mRNA | AK012132.1 | - | 5 - 6 | 1278 | 97 | |
| AK028787.1 | - | 5 - 6 | 1010 | 97 | ||
| AK034503.1 | - | 4 - 5 | 901 | 97 | ||
| AK166083.1 | - | 5 - 6 | 1014 | 97 | ||
| BC099950.1 | - | 4 - 5 | 855 | 30094307 | 97 | |
| BC138218.1 | - | 3 - 4 | 784 | 8861237 | 97 | |
| BC145204.1 | - | 3 - 4 | 784 | 9053755 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dzip1l
Genomic Map