Assay Details
| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028846.5 | NP_083122.1 | 9 - 10 | 851 | 86 | |
| GenBank mRNA | AF449715.1 | - | 9 - 10 | 802 | 86 | |
| AK029890.1 | - | 748 | 86 | |||
| AK047242.1 | - | 9 - 10 | 770 | 86 | ||
| AK054279.1 | - | 9 - 10 | 794 | 86 | ||
| AK087412.1 | - | 9 - 10 | 762 | 86 | ||
| AK163663.1 | - | 9 - 10 | 761 | 86 | ||
| AK173083.1 | - | 9 - 10 | 753 | 86 | ||
| BC037199.1 | - | 3 - 4 | 235 | 86 | ||
| BC079674.1 | - | 740 | 30531994 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp20
Genomic Map