Assay Details
| Gene Symbol: Ccdc181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029115.3 | NP_083391.2 | 3 - 4 | 1249 | 109 | |
| GenBank mRNA | AK076640.1 | - | 3 - 4 | 1244 | 109 | |
| AK149168.1 | - | 3 - 4 | 1244 | 109 | ||
| BC048086.1 | - | 3 - 4 | 1141 | 6390120 | 109 | |
| BC060044.1 | - | 3 - 4 | 1248 | 109 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc181
Genomic Map