Assay Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290649.1 | NP_001277578.1 | 17 - 18 | 1993 | 125 | |
| NM_029793.2 | NP_084069.1 | 18 - 19 | 2068 | 125 | ||
| NR_110968.1 | 18 - 19 | 2068 | 125 | |||
| GenBank mRNA | AK028516.1 | - | 18 - 19 | 1988 | 125 | |
| AK044987.1 | - | 17 - 18 | 2478 | 125 | ||
| BC037037.1 | - | 13 - 14 | 1404 | 125 | ||
| BC043090.1 | - | 18 - 19 | 1933 | 6418353 | 125 | |
| BC053068.1 | - | 17 - 18 | 1921 | 6405595 | 125 | |
| BC080733.1 | - | 17 - 18 | 1993 | 30110235 | 125 | |
More Information
Set Membership: |
|

Golga1
Genomic Map