Assay Details
| Gene Symbol: Golga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290649.1 | NP_001277578.1 | 6 - 7 | 691 | 80 | |
| NM_029793.2 | NP_084069.1 | 7 - 8 | 766 | 80 | ||
| NR_110968.1 | 7 - 8 | 766 | 80 | |||
| GenBank mRNA | AK002954.1 | - | 7 - 8 | 710 | 80 | |
| AK008972.1 | - | 7 - 8 | 665 | 80 | ||
| AK028516.1 | - | 7 - 8 | 685 | 80 | ||
| AK044987.1 | - | 7 - 8 | 714 | 80 | ||
| BC024637.1 | - | 7 - 8 | 688 | 80 | ||
| BC037037.1 | - | 3 - 4 | 203 | 80 | ||
| BC043090.1 | - | 7 - 8 | 631 | 6418353 | 80 | |
| BC053068.1 | - | 6 - 7 | 619 | 6405595 | 80 | |
| BC080733.1 | - | 6 - 7 | 691 | 30110235 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Golga1
Genomic Map