Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 21 - 22 | 2629 | 62 | |
| NM_138306.2 | NP_612179.2 | 20 - 21 | 2028 | 62 | ||
| GenBank mRNA | AK087503.1 | - | 21 - 22 | 2631 | 62 | |
| AK170936.1 | - | 17 - 18 | 1651 | 62 | ||
| BC014860.1 | - | 5 - 6 | 495 | 62 | ||
| BC049228.1 | - | 20 - 21 | 2064 | 3668704 | 62 | |
| CD564532.1 | - | 5 - 6 | 510 | 62 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map