Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 25 - 26 | 2920 | 59 | |
| NM_138306.2 | NP_612179.2 | 24 - 25 | 2319 | 59 | ||
| GenBank mRNA | AK170936.1 | - | 21 - 22 | 1942 | 59 | |
| AK204403.1 | - | 4 - 5 | 248 | 59 | ||
| BC014860.1 | - | 9 - 10 | 789 | 59 | ||
| BC049228.1 | - | 24 - 25 | 2355 | 3668704 | 59 | |
| BE862117.1 | - | 1 - 2 | 48 | 59 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK088841.1 | - | Chr. - - [-] on Build | 59 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map