Assay Details
| Gene Symbol: Prr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145589.2 | NP_663564.2 | 9 - 10 | 1829 | 109 | |
| GenBank mRNA | AK133884.1 | - | 9 - 10 | 1793 | 109 | |
| AK139559.1 | - | 10 - 11 | 1604 | 109 | ||
| AK145816.1 | - | 10 - 11 | 1609 | 109 | ||
| AK151203.1 | - | 10 - 11 | 1661 | 109 | ||
| AK182875.1 | - | 3 - 4 | 268 | 109 | ||
| AK207496.1 | - | 2 - 3 | 186 | 109 | ||
| BC055033.1 | - | 9 - 10 | 1829 | 6310272 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prr14
Genomic Map