Assay Details
| Gene Symbol: Hadhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289798.1 | NP_001276727.1 | 3 - 4 | 251 | 102 | |
| NM_001289799.1 | NP_001276728.1 | 3 - 4 | 247 | 102 | ||
| NM_145558.2 | NP_663533.1 | 3 - 4 | 204 | 102 | ||
| GenBank mRNA | AK033462.1 | - | 3 - 4 | 186 | 102 | |
| AK076814.1 | - | 3 - 4 | 167 | 102 | ||
| AK083164.1 | - | 3 - 4 | 212 | 102 | ||
| AK083767.1 | - | 3 - 4 | 167 | 102 | ||
| AK150889.1 | - | 3 - 4 | 166 | 102 | ||
| AK168594.1 | - | 3 - 4 | 168 | 102 | ||
| AK169637.1 | - | 3 - 4 | 180 | 102 | ||
| BC005585.1 | - | 2 - 3 | 135 | 3158015 | 102 | |
| BY337615.1 | - | 3 - 4 | 205 | 102 | ||
| BY778233.1 | - | 3 - 4 | 181 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hadhb
Genomic Map