Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 9 - 10 | 1355 | 77 | |
| GenBank mRNA | CF538789.1 | - | 4 - 5 | 341 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map