Assay Details
| Gene Symbol: Sod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013671.3 | NP_038699.2 | 3 - 4 | 545 | 64 | |
| GenBank mRNA | AK002428.1 | - | 3 - 4 | 405 | 64 | |
| AK002534.1 | - | 3 - 4 | 406 | 64 | ||
| AK012354.1 | - | 3 - 4 | 403 | 64 | ||
| AK135435.1 | - | 3 - 4 | 534 | 64 | ||
| AK152047.1 | - | 3 - 4 | 403 | 64 | ||
| AK167521.1 | - | 3 - 4 | 406 | 64 | ||
| AK168082.1 | - | 3 - 4 | 406 | 64 | ||
| AK168591.1 | - | 3 - 4 | 537 | 64 | ||
| AK179113.1 | - | 2 - 3 | 165 | 64 | ||
| BC010548.1 | - | 3 - 4 | 381 | 3581909 | 64 | |
| BC018173.1 | - | 2 - 3 | 326 | 64 | ||
| CT010273.1 | - | 3 - 4 | 338 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sod2
Genomic Map