Assay Details
| Gene Symbol: Lhfpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177763.3 | NP_808431.2 | 1 - 2 | 173 | 128 | |
| GenBank mRNA | AK220449.1 | - | 1 - 2 | 117 | 128 | |
| BC085130.1 | - | 1 - 2 | 120 | 5704741 | 128 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC085130.1,NM_177763.3 transcripts.
This assay is in the 5'UTR region of the BC085130.1,NM_177763.3 transcripts.
Set Membership: |
|

Lhfpl4
Genomic Map