Assay Details
| Gene Symbol: Lrrc57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025657.3 | NP_079933.2 | 1 - 2 | 124 | 75 | |
| GenBank mRNA | BC034894.1 | - | 1 - 2 | 83 | 3328402 | 75 |
| BU604191.1 | - | 1 - 2 | 127 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC034894.1, NM_025657.3 transcripts.
Set Membership: |
|

Lrrc57
Genomic Map