Assay Details
| Gene Symbol: Cpa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027926.2 | NP_082202.1 | 1 - 2 | 89 | 115 | |
| GenBank mRNA | AK030705.1 | - | 1 - 2 | 89 | 115 | |
| BC061206.1 | - | 1 - 2 | 96 | 30249166 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030705.1, BC061206.1, NM_027926.2 transcripts.
Set Membership: |
|

Cpa4
Genomic Map