Assay Details
| Gene Symbol: Ppm1j | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027982.2 | NP_082258.2 | 6 - 7 | 1081 | 82 | |
| GenBank mRNA | AK009235.1 | - | 6 - 7 | 1020 | 82 | |
| AY184802.1 | - | 6 - 7 | 1081 | 82 | ||
| BC117498.1 | - | 6 - 7 | 1077 | 40106734 | 82 | |
| BC121788.1 | - | 6 - 7 | 1076 | 40106732 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppm1j
Genomic Map