Assay Details
| Gene Symbol: Gorasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028976.2 | NP_083252.1 | 4 - 5 | 468 | 118 | |
| GenBank mRNA | AK154986.1 | - | 4 - 5 | 445 | 118 | |
| AK170627.1 | - | 4 - 5 | 432 | 118 | ||
| BC012251.1 | - | 4 - 5 | 468 | 4166619 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gorasp1
Genomic Map