Assay Details
| Gene Symbol: Scaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134123.3 | NP_598884.2 | 17 - 18 | 2488 | 97 | |
| GenBank mRNA | AK032418.1 | - | 690 | 97 | ||
| AK122451.1 | - | 17 - 18 | 2355 | 97 | ||
| AK161113.1 | - | 3 - 4 | 400 | 97 | ||
| BC030846.1 | - | 4 - 5 | 486 | 97 | ||
| BC038363.1 | - | 17 - 18 | 2132 | 5371946 | 97 | |
| BC075621.1 | - | 17 - 18 | 2481 | 6406441 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scaf8
Genomic Map