Assay Details
| Gene Symbol: Crtc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173863.2 | NP_776288.2 | 8 - 9 | 841 | 81 | |
| GenBank mRNA | AK144840.1 | - | 8 - 9 | 842 | 82 | |
| BC011210.1 | - | 8 - 9 | 680 | 4195538 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC011210.1 transcripts.
Set Membership: |
|

Crtc3
Genomic Map