Assay Details
| Gene Symbol: Rcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173867.5 | NP_776292.1 | 11 - 12 | 1569 | 63 | |
| GenBank mRNA | AK076040.1 | - | 11 - 12 | 1500 | 63 | |
| AK076046.1 | - | 1800 | 63 | |||
| AK129370.1 | - | 10 - 11 | 1273 | 63 | ||
| BC020385.1 | - | 9 - 10 | 1102 | 63 | ||
| BC034759.1 | - | 8 - 9 | 889 | 63 | ||
| BC060624.1 | - | 11 - 12 | 62 | |||
| BC086666.1 | - | 11 - 12 | 1492 | 30634817 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rcc2
Genomic Map