Assay Details
| Gene Symbol: Samd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175021.3 | NP_778186.2 | 7 - 8 | 1684 | 87 | |
| GenBank mRNA | BC042901.1 | - | 7 - 8 | 1737 | 5132736 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|

Samd4b
Genomic Map