Assay Details
| Gene Symbol: P3h4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176830.2 | NP_789800.1 | 4 - 5 | 1092 | 91 | |
| GenBank mRNA | AK029271.1 | - | 4 - 5 | 972 | 91 | |
| BC031856.1 | - | 4 - 5 | 1092 | 3968941 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

P3h4
Genomic Map