Assay Details
| Gene Symbol: Ubash3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176860.5 | NP_789830.1 | 12 - 13 | 1742 | 109 | |
| GenBank mRNA | AB075602.1 | - | 12 - 13 | 1737 | 109 | |
| AK013361.1 | - | 12 - 13 | 1810 | 109 | ||
| AK034450.1 | - | 12 - 13 | 1933 | 109 | ||
| AK035764.1 | - | 12 - 13 | 1743 | 109 | ||
| AK133948.1 | - | 12 - 13 | 1740 | 109 | ||
| AK150909.1 | - | 12 - 13 | 2309 | 109 | ||
| AK151721.1 | - | 12 - 13 | 2310 | 109 | ||
| AK151895.1 | - | 12 - 13 | 1943 | 109 | ||
| AK152013.1 | - | 12 - 13 | 1943 | 109 | ||
| AK154576.1 | - | 12 - 13 | 1964 | 109 | ||
| BC053436.1 | - | 12 - 13 | 1714 | 6510702 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubash3b
Genomic Map