Assay Details
| Gene Symbol: Suds3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122666.2 | NP_001116138.1 | 5 - 6 | 544 | 78 | |
| NM_178622.5 | NP_848737.3 | 5 - 6 | 544 | 78 | ||
| GenBank mRNA | AF469109.1 | - | 5 - 6 | 484 | 78 | |
| AK010274.1 | - | 5 - 6 | 447 | 78 | ||
| AK043652.1 | - | 5 - 6 | 487 | 78 | ||
| AK045475.1 | - | 5 - 6 | 498 | 78 | ||
| AK138947.1 | - | 5 - 6 | 495 | 78 | ||
| AK150633.1 | - | 5 - 6 | 482 | 78 | ||
| BC040088.1 | - | 3 - 4 | 157 | 78 | ||
| BC055764.1 | - | 5 - 6 | 497 | 6402735 | 78 | |
| BC062176.1 | - | 5 - 6 | 411 | 6514171 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Suds3
Genomic Map