Assay Details
| Gene Symbol: Suds3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122666.2 | NP_001116138.1 | 9 - 10 | 879 | 64 | |
| NM_178622.5 | NP_848737.3 | 9 - 10 | 879 | 64 | ||
| GenBank mRNA | AF469109.1 | - | 9 - 10 | 819 | 64 | |
| AK010274.1 | - | 9 - 10 | 783 | 64 | ||
| AK043652.1 | - | 9 - 10 | 822 | 64 | ||
| AK045475.1 | - | 9 - 10 | 833 | 64 | ||
| AK138947.1 | - | 9 - 10 | 830 | 64 | ||
| AK150633.1 | - | 9 - 10 | 817 | 64 | ||
| BC036562.1 | - | 4 - 5 | 252 | 64 | ||
| BC040088.1 | - | 7 - 8 | 492 | 64 | ||
| BC055764.1 | - | 9 - 10 | 832 | 6402735 | 64 | |
| BC062176.1 | - | 9 - 10 | 746 | 6514171 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Suds3
Genomic Map