Assay Details
| Gene Symbol: Poldip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178627.3 | NP_848742.1 | 1 - 2 | 140 | 81 | |
| GenBank mRNA | AK052722.1 | - | 1 - 2 | 81 | ||
| AK081139.1 | - | 1 - 2 | 100 | 81 | ||
| AK083924.1 | - | 1 - 2 | 141 | 81 | ||
| AK087446.1 | - | 1 - 2 | 149 | 81 | ||
| AK150130.1 | - | 1 - 2 | 118 | 81 | ||
| AK152060.1 | - | 1 - 2 | 181 | 81 | ||
| AK167552.1 | - | 1 - 2 | 84 | 81 | ||
| AK220221.1 | - | 1 - 2 | 156 | 81 | ||
| BC037652.1 | - | 1 - 2 | 125 | 5136112 | 81 | |
| BC046505.1 | - | 1 - 2 | 69 | 4912042 | 81 | |
| BC058225.1 | - | 1 - 2 | 136 | 6307930 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poldip3
Genomic Map