Assay Details
| Gene Symbol: Poldip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178627.3 | NP_848742.1 | 5 - 6 | 894 | 86 | |
| GenBank mRNA | AK081139.1 | - | 5 - 6 | 854 | 86 | |
| AK083924.1 | - | 5 - 6 | 895 | 86 | ||
| AK087446.1 | - | 5 - 6 | 903 | 86 | ||
| AK088088.1 | - | 1 - 2 | 116 | 86 | ||
| AK150130.1 | - | 4 - 5 | 785 | 86 | ||
| AK152060.1 | - | 5 - 6 | 935 | 86 | ||
| AK167552.1 | - | 5 - 6 | 838 | 86 | ||
| AK220221.1 | - | 5 - 6 | 910 | 86 | ||
| BC037652.1 | - | 5 - 6 | 879 | 5136112 | 86 | |
| BC046505.1 | - | 5 - 6 | 823 | 4912042 | 86 | |
| BC058225.1 | - | 5 - 6 | 890 | 6307930 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poldip3
Genomic Map