Assay Details
| Gene Symbol: Poldip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178627.3 | NP_848742.1 | 6 - 7 | 977 | 96 | |
| GenBank mRNA | AK081139.1 | - | 6 - 7 | 937 | 96 | |
| AK083924.1 | - | 6 - 7 | 978 | 96 | ||
| AK087446.1 | - | 6 - 7 | 986 | 96 | ||
| AK088088.1 | - | 2 - 3 | 199 | 96 | ||
| AK152060.1 | - | 6 - 7 | 1018 | 96 | ||
| AK167552.1 | - | 6 - 7 | 921 | 96 | ||
| AK220221.1 | - | 6 - 7 | 993 | 96 | ||
| BC037652.1 | - | 6 - 7 | 962 | 5136112 | 96 | |
| BC046505.1 | - | 6 - 7 | 906 | 4912042 | 96 | |
| BC058225.1 | - | 6 - 7 | 973 | 6307930 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Poldip3
Genomic Map