Assay Details
| Gene Symbol: Blmh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178645.4 | NP_848760.1 | 4 - 5 | 685 | 80 | |
| GenBank mRNA | AK029274.1 | - | 3 - 4 | 1578 | 80 | |
| AK035228.1 | - | 4 - 5 | 559 | 80 | ||
| AK040799.1 | - | 4 - 5 | 575 | 80 | ||
| AK049461.1 | - | 4 - 5 | 597 | 80 | ||
| AK051441.1 | - | 4 - 5 | 584 | 80 | ||
| AK053872.1 | - | 4 - 5 | 565 | 80 | ||
| AK150438.1 | - | 4 - 5 | 584 | 80 | ||
| AK167325.1 | - | 4 - 5 | 565 | 80 | ||
| BC027037.1 | - | 4 - 5 | 541 | 5364667 | 80 | |
| BC027362.1 | - | 4 - 5 | 574 | 4935208 | 80 | |
| BC027403.1 | - | 4 - 5 | 595 | 4952098 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Blmh
Genomic Map