Assay Details
| Gene Symbol: Mief1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178719.5 | NP_848834.2 | 1 - 2 | 98 | 62 | |
| GenBank mRNA | AK038466.1 | - | 1 - 2 | 71 | 62 | |
| AK041184.1 | - | 1 - 2 | 77 | 62 | ||
| AK050546.1 | - | 1 - 2 | 59 | 62 | ||
| AK080375.1 | - | 1 - 2 | 44 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK038466.1, AK050546.1, AK080375.1, NM_178719.5 transcripts.
Set Membership: |
|

Mief1
Genomic Map