Assay Details
| Gene Symbol: Tiparp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178892.5 | NP_849223.2 | 3 - 4 | 1312 | 74 | |
| GenBank mRNA | AK028529.1 | - | 3 - 4 | 1313 | 74 | |
| AK032453.1 | - | 3 - 4 | 1296 | 74 | ||
| AK150248.1 | - | 3 - 4 | 1294 | 74 | ||
| AK162094.1 | - | 3 - 4 | 1318 | 74 | ||
| AK163347.1 | - | 3 - 4 | 1352 | 74 | ||
| BC003919.1 | - | 2 - 3 | 572 | 74 | ||
| BC068173.1 | - | 3 - 4 | 1286 | 30616032 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tiparp
Genomic Map