Assay Details
| Gene Symbol: Spata7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289572.1 | NP_001276501.1 | 9 - 10 | 1202 | 136 | |
| NM_001289573.1 | NP_001276502.1 | 8 - 9 | 797 | 136 | ||
| NM_001289574.1 | NP_001276503.1 | 10 - 11 | 1364 | 136 | ||
| NM_178914.4 | NP_849245.2 | 10 - 11 | 1298 | 136 | ||
| GenBank mRNA | AK036359.1 | - | 10 - 11 | 1282 | 136 | |
| AK077500.1 | - | 10 - 11 | 1209 | 136 | ||
| AK153698.1 | - | 8 - 9 | 773 | 136 | ||
| AK161308.1 | - | 9 - 10 | 1148 | 136 | ||
| BC046960.1 | - | 10 - 11 | 1256 | 6391450 | 136 | |
| DQ196101.1 | - | 9 - 10 | 1028 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spata7
Genomic Map