Assay Details
| Gene Symbol: Nostrin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181547.3 | NP_853525.3 | 4 - 5 | 347 | 68 | |
| GenBank mRNA | AJ564264.1 | - | 4 - 5 | 257 | 68 | |
| AK052926.1 | - | 4 - 5 | 339 | 68 | ||
| AY280963.1 | - | 4 - 5 | 300 | 68 | ||
| BC069942.1 | - | 4 - 5 | 324 | 30443942 | 68 | |
| BY061165.1 | - | 4 - 5 | 347 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nostrin
Genomic Map