Assay Details
| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181729.2 | NP_859418.1 | 2524 | 64 | ||
| GenBank mRNA | AJ010752.1 | - | 2 - 3 | 148 | 64 | |
| BC120907.2 | - | 19 - 20 | 2412 | 40087870 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Muc6
Genomic Map