Assay Details
| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181729.2 | NP_859418.1 | 3930 | 93 | ||
| GenBank mRNA | BC120907.2 | - | 29 - 30 | 3818 | 40087870 | 93 |
More Information
Updated Mapping Information
Set Membership: |
|

Muc6
Genomic Map