Assay Details
| Gene Symbol: Rbmx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166623.1 | NP_001160095.1 | 5 - 6 | 759 | 78 | |
| NM_011252.4 | NP_035382.1 | 5 - 6 | 756 | 78 | ||
| NR_029425.1 | 5 - 6 | 756 | 78 | |||
| GenBank mRNA | AJ237846.1 | - | 5 - 6 | 686 | 78 | |
| AJ237847.1 | - | 5 - 6 | 701 | 78 | ||
| AK083899.1 | - | 5 - 6 | 757 | 78 | ||
| AK135164.1 | - | 5 - 6 | 761 | 78 | ||
| AK165006.1 | - | 5 - 6 | 702 | 78 | ||
| AK178027.1 | - | 1 - 2 | 98 | 78 | ||
| BC003710.1 | - | 5 - 6 | 754 | 3599678 | 78 | |
| CK619886.1 | - | 4 - 5 | 562 | 78 | ||
More Information
Set Membership: |
|

Rbmx
Genomic Map