Assay Details
| Gene Symbol: Ccdc126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175098.5 | NP_780307.1 | 1 - 2 | 128 | 96 | |
| GenBank mRNA | AK018135.1 | - | 1 - 2 | 129 | 96 | |
| AK159264.1 | - | 1 - 2 | 96 | |||
| BC117837.2 | - | 1 - 2 | 76 | 40093139 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018135.1, AK159264.1, BC117837.2, NM_175098.5 transcripts.
Set Membership: |
|

Ccdc126
Genomic Map