Assay Details
| Gene Symbol: Hnrnpa2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_104468.1 | 9 - 10 | 1149 | 81 | ||
| GenBank mRNA | AK089012.1 | - | 9 - 10 | 1100 | 81 | |
| BC141253.1 | - | 9 - 10 | 983 | 9055865 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hnrnpa2b1
Genomic Map