Assay Details
| Gene Symbol: Frs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177798.3 | NP_808466.1 | 7 - 8 | 611 | 58 | |
| GenBank mRNA | AK028813.1 | - | 7 - 8 | 627 | 58 | |
| BC043109.1 | - | 7 - 8 | 611 | 6406912 | 58 | |
| BC055334.1 | - | 7 - 8 | 647 | 6830555 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Frs2
Genomic Map