Assay Details
| Gene Symbol: Kir3dl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177749.3 | NP_808417.2 | 7 - 8 | 1140 | 148 | |
| GenBank mRNA | AY130461.1 | - | 7 - 8 | 1126 | 149 | |
| AY530782.1 | - | 7 - 8 | 1113 | 148 | ||
| BC141012.1 | - | 7 - 8 | 1126 | 9055624 | 148 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036490 | BC141012.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036490 | AY530782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036490 | NM_177749.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036490 | AY130461.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995617 | AY530783.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995617 | AY530782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995617 | NM_177749.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1995617 | AY130461.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717201 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 856753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254798 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507331 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119323 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145422 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P174340 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036490 | BC141012.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036490 | AY530782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036490 | NM_177749.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036490 | AY130461.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995617 | AY530783.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995617 | AY530782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995617 | NM_177749.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1995617 | AY130461.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32681236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32681228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32681251 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09430171 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P174340 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09430199 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03759875 | - | - |
| Whole Mouse Genome G4122A | A_52_P538451 | AY530783.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P538451 | BC141012.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P538451 | AY530782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P538451 | NM_177749.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P538451 | AY130461.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09430199 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P174340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681306 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32681303 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681225 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681250 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14823789 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32681266 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10601870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601869 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10601872 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10601867 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00008353.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008353.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008352.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008352.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008350.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008350.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008357.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008357.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008359.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008359.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008360.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008360.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008354.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008354.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008351.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008351.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008355.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008355.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008356.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008356.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00008358.mm.1 | NM_177749.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00008358.mm.1 | TC0X00001331.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 725005 | - | - |
| ABI Mouse Genome Survey Microarray | 877969 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4060280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4060280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4920685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004920685 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004060280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734355 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1250087 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232657 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232657 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250087 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2734355 | - | - |
| mouse-6 v1.1 (Illumina) | scl0002941.1_145-S | - | - |
| mouse-6 v1.1 (Illumina) | scl00245616.2_11-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4060280 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4920685 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00245616.2_11-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002941.1_145-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4920685 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4060280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1250087 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2734355 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250087 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734355 | - | - |

Kir3dl1
Genomic Map