Assay Details
| Gene Symbol: Rrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK156199.1 | - | 1 - 2 | 118 | 115 | |
| BC014703.1 | - | 1 - 2 | 126 | 4237895 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrp9
Genomic Map