Assay Details
| Gene Symbol: Vamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080557.1 | NP_001074026.1 | 1 - 2 | 151 | 58 | |
| NM_009496.3 | NP_033522.1 | 1 - 2 | 151 | 58 | ||
| GenBank mRNA | AK018783.1 | - | 1 - 2 | 151 | 58 | |
| AK037711.1 | - | 1 - 2 | 152 | 58 | ||
| AK078156.1 | - | 1 - 2 | 152 | 58 | ||
| BC049902.1 | - | 1 - 2 | 113 | 4500872 | 58 | |
| BC057587.1 | - | 1 - 2 | 130 | 6415847 | 58 | |
| U61751.1 | - | 1 - 2 | 97 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK018783.1, AK037711.1, AK078156.1, BC049902.1, BC057587.1, NM_001080557.1, NM_009496.3, U61751.1 transcripts.
Set Membership: |
|

Vamp1
Genomic Map