Assay Details
| Gene Symbol: Hnrnpdl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016690.4 | NP_057899.2 | 2 - 3 | 667 | 91 | |
| GenBank mRNA | AB017020.1 | - | 2 - 3 | 373 | 91 | |
| AK011581.1 | - | 2 - 3 | 356 | 91 | ||
| AK011767.1 | - | 2 - 3 | 356 | 91 | ||
| AK172629.1 | - | 2 - 3 | 326 | 91 | ||
| BC021374.1 | - | 2 - 3 | 325 | 5039077 | 91 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Hnrnpdl
Genomic Map