Assay Details
| Gene Symbol: Rbpms2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028030.3 | NP_082306.2 | 5 - 6 | 647 | 71 | |
| GenBank mRNA | AK144659.1 | - | 5 - 6 | 672 | 71 | |
| AK156633.1 | - | 5 - 6 | 647 | 71 | ||
| BC021788.1 | - | 5 - 6 | 645 | 4236661 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rbpms2
Genomic Map