Assay Details
| Gene Symbol: Ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025348.2 | NP_079624.1 | 2 - 3 | 114 | 92 | |
| GenBank mRNA | AK003133.1 | - | 2 - 3 | 105 | 92 | |
| AK019099.1 | - | 2 - 3 | 116 | 92 | ||
| BC061002.1 | - | 107 | 92 | |||
| BC120712.1 | - | 2 - 3 | 108 | 40129635 | 92 | |
| BC120714.1 | - | 2 - 3 | 108 | 40129637 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ndufa3
Genomic Map