| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0X00002640.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002640.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002628.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002628.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002630.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002630.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002638.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002627.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002627.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002641.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002641.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002626.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002626.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002625.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002625.mm.1 |
TC0X00000396.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0X00002642.mm.1 |
NM_009688.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0X00002642.mm.1 |
TC0X00000396.mm.1 |
MTA TC |