| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700017388.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017388.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017388.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017374.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017374.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017374.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017378.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017378.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017378.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017381.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017381.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017381.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017375.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017375.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017375.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017383.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017383.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017383.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017392.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017392.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017392.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017386.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017386.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017386.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017397.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017397.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017397.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017376.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017376.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017376.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017382.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017382.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017382.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017384.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017384.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017384.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017387.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017387.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017387.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017379.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017379.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017379.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017380.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017380.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017380.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017389.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017389.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017389.mm.1 |
TC1700001876.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700017391.mm.1 |
NM_008997.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017391.mm.1 |
NM_008997.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700017391.mm.1 |
TC1700001876.mm.1 |
MTA TC |